Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specificity: ABS 051-04-02 binds human PYY (3-36). This antibody does not cross-react with PYY (1-36) from mouse, rat, dog or pig samples.
Reactivity: ABS 051-04-02 binds free PYY (3-36) in solution. The recommended ELISA dilution is 1:20,000 for PYY (3-36) coated directly at 1 µg/mL. ABS 051-04-02 can be used as a capture antibody in a sandwich ELISA using biotinylated detection antibody ABS 029-01-02.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Peptide tyrosine-tyrosine amide (3-36), or PYY (3-36), is a major metabolite of the gut hormone PYY and is produced by the action of dipeptidyl peptidase IV on PYY in the intestinal brush border and in the circulation. PYY (3-36) is a potent inhibitor of food intake in rats and humans, acting selectively on the Y2 receptor to inhibit orexigenic NPY neurons in the arcuate nucleus, thus disinhibiting anorexigenic POMC neurons. The anorexigenic effect of PYY (3-36) is additive to that of the co-secreted gut hormone GLP-1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Peptide tyrosine tyrosine; Peptide YY; Peptide YY3-36; Peptide YY3-37; prepro-PYY; PYY; PYY II; PYY-1; PYY-2; PYY-I; PYYI; PYYII
Gene Aliases: PYY; PYY-I; PYY1
UniProt ID: (Human) P10082
Entrez Gene ID: (Human) 5697
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support